CAS RN 1302-78-9
IUPAC Name:
tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
Synonyms:
tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
Molecular Weight (g/mol):
360.31
Molecular Formula:
Al2H2O12Si4
InChi Key:
KMDMICAGCNGGEB-UHFFFAOYSA-N
SMILES:
O.[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Si+4].[Si+4].[Si+4].[Si+4]
1
–
5
of
5
results
1
Humidity Sponge™ Dessicant Window Type, Fisherbrand™
Meet all drying requirements with these humidity sponges. Fisherbrand™ Dessicant Window Type Humidity Sponges™ are non-toxic and Class M3.5 cleanroom-compatible.
2
Humidity Sponge™, Fisherbrand™
Humidity Sponge Dessicant; Fisherbrand; Reusable Type
3
Bentonite
CAS: 1302-78-9
4
Bentonite, sodium form
CAS: 1302-78-9 MDL Number: MFCD00130611
5
Promotions Available
Bentonite, Powder, NF, Spectrum™ Chemical
Bentonite, Powder, NF, Spectrum™ Chemical
CAS: 1302-78-9 Molecular Formula: Al2H2O12Si4 Molecular Weight (g/mol): 360.31 InChI Key: KMDMICAGCNGGEB-UHFFFAOYSA-N IUPAC Name: tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide SMILES: O.[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Si+4].[Si+4].[Si+4].[Si+4]