missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, Powder, NF, Spectrum™ Chemical

Catalog No. 18600871
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
22.5 kg
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600871 22.5 kg Fiber Drum
18600868 500 g Fiber Drum
18600869 2.5 kg Fiber Drum
18600870 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600871 Supplier Spectrum Chemical Mfg Cor Supplier No. BE12522.5KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BE125, 1302-78-9

Spectrum™ Chemical Bentonite, Powder, NF is used as suspending and emulsifying agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1302-78-9
Molecular Formula Al2H2O12Si4
Molecular Weight (g/mol) 360.31
InChI Key KMDMICAGCNGGEB-UHFFFAOYSA-N
IUPAC Name tetrasilicon(4+) dialuminium(3+) hydrate undecaoxidandiide
SMILES O.[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Si+4].[Si+4].[Si+4].[Si+4]

Specifications

CAS Max % 100%
Loss on Drying 5.0-8%
Packaging Fiber Drum
Quantity 22.5 kg
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Bentonite, Powder