missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, sodium form

Catalog No. AAA1579536
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1579536 500 g
AAA157950E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1579536 Supplier Thermo Scientific Chemicals Supplier No. A1579536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Forms a colloidal suspension in water | CAS: 1302-78-9

Bentonite, sodium form is used as a suspending agent, a gel binder, a filler in ceramics and a catalyst support.Sodium bentonite is widely utilized as a natural sealant in recreational ponds, dairy and sewage lagoons and city landfills. Due to its colloidal property, it is employed in drilling mud for oil and gas wells. Due to its swelling property, it provides a self-sealing low permeability barrier. It combines with polymers and enhances the surface modifications, which are used for rheological or sealing performance in geo-environmental applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bentonite, sodium form is used as a suspending agent, a gel binder, a filler in ceramics and a catalyst support.Sodium bentonite is widely utilized as a natural sealant in recreational ponds, dairy and sewage lagoons and city landfills. Due to its colloidal property, it is employed in drilling mud for oil and gas wells. Due to its swelling property, it provides a self-sealing low permeability barrier. It combines with polymers and enhances the surface modifications, which are used for rheological or sealing performance in geo-environmental applications.

Solubility
Insoluble in water and forms a colloidal solution.

Notes
Forms a colloidal suspension in water.

Chemical Identifiers

CAS 1302-78-9
MDL Number MFCD00130611

Specifications

Quantity 500 g
Solubility Information Insoluble in water and forms a colloidal solution.
Chemical Name or Material Bentonite
EINECSNumber 215-108-5
RTECSNumber CT9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only