missing translation for 'onlineSavingsMsg'
Learn More

Humidity Sponge™, Fisherbrand™

Catalog No. 075801
Change view
Click to view available options
Chemical Name or Material:
Fisherbrand™ Humidity Sponge™ Dessicant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
075801 Fisherbrand™ Humidity Sponge™ Dessicant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 075801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Humidity Sponge Dessicant; Fisherbrand; Reusable Type

  • Non-toxic desiccant
  • Reusable
  • Material is made of DuPont's Tyvek™ breathable, high-density polyethylene
  • Perfect for drugs, food, electronics, optics and equipment
  • Bags are guaranteed to retain 16% moisture by weight, typical results are 37% moisture absorption
  • Desiccant meets US FDA requirements
  • Certified ISO Class 5 (Class M3.5) cleanroom compatible

Specifications

Chemical Name or Material Fisherbrand™ Humidity Sponge™ Dessicant
CAS 1302-78-9
Material HDPE
Quantity 40 Pk.
Grade Certified
Weight 11 g