missing translation for 'onlineSavingsMsg'
Learn More

Humidity Sponge™ Dessicant Window Type, Fisherbrand™

Catalog No. 07580
Change view
Click to view available options
Chemical Name or Material:
Fisherbrand™ Humidity Sponge™ Dessicant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
07580 Fisherbrand™ Humidity Sponge™ Dessicant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07580
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meet all drying requirements with these humidity sponges. Fisherbrand™ Dessicant Window Type Humidity Sponges™ are non-toxic and Class M3.5 cleanroom-compatible.

  • Bags result in 37% moisture absorption
  • Indicators show when moisture capacity has been reached
  • One bag will retain 16% moisture in a container with a volume of two gallons (476 cu. in.)
  • Bag size: 7.6 x 7.6 x 0.6 cm (3 x 3 x 1/4 in.)

Specifications

Chemical Name or Material Fisherbrand™ Humidity Sponge™ Dessicant
CAS 1302-78-9
Material Sodium Calcium Aluminosilicate Hydrate
Quantity 40 Pk.
Grade Certified
Weight Each bag weighs 10 g (0.35 oz.)