missing translation for 'onlineSavingsMsg'
Learn More

Bentonite

Catalog No. AC447445000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447445000 500 g Plastic bottle
AC447440025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447445000 Supplier Thermo Scientific Chemicals Supplier No. 447445000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1302-78-9

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent, Lewis acid

Chemical Identifiers

CAS 1302-78-9

Specifications

Packaging Plastic bottle
Quantity 500 g
Merck Index 15,1057
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in acids and common organic solvents
Chemical Name or Material Bentonite
EINECSNumber 215-108-5

RUO – Research Use Only