accessibility menu, dialog, popup

UserName

OHIClN

CAS RN 130-26-7

IUPAC Name: 5-chloro-7-iodoquinolin-8-ol
Synonyms: clioquinol caswell no. 193 iodochloroxychinolinum jodchloroxychinolinum racet dioquinol ala-quin emaform 5 chloro 7 iodo 8 quinolinol enterovalodon
Molecular Weight (g/mol): 305.5
Molecular Formula: C9H5ClINO
InChi Key: QCDFBFJGMNKBDO-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Cl)C=C1I
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)