missing translation for 'onlineSavingsMsg'
Learn More

Clioquinol, Approx. 95%, Spectrum™ Chemical

Catalog No. 18609822
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609822 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18609822 Supplier Spectrum Chemical Mfg Cor Supplier No. CL109100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 130-26-7
Molecular Weight (g/mol) 178.5

Specifications

CAS Max % 1
Boiling Point 350.4°C (662.72°F)
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity ∼95%
Grade Ungraded
Physical Form Powder or Waxy Solid
Chemical Name or Material Clioquinol

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.