accessibility menu, dialog, popup

UserName

CH3OH3COOOHH3COONO(E)

CAS RN 128794-94-5

IUPAC Name: 2-(morpholin-4-yl)ethyl (4E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydro-2-benzofuran-5-yl)-4-methylhex-4-enoate
Synonyms: mycophenolate mofetil munoloc MMF mycophenolic acid morpholinoethyl ester cellcept CellCept mycophenolate mofetil preparation mycophenylate mofetil mycophenolate mofetil (substance) mycophenolate mofetil (product)
Molecular Weight (g/mol): 433.5
Molecular Formula: C23H31NO7
InChi Key: RTGDFNSFWBGLEC-SYZQJQIISA-N
SMILES: COC1=C(C)C2=C(C(=O)OC2)C(O)=C1C\C=C(/C)CCC(=O)OCCN1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)