missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Mofetil 98.0+%, TCI America™

Catalog No. M23871G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23871G 1 g
M2387100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23871G Supplier TCI America Supplier No. M23871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128794-94-5
Molecular Formula C23H31NO7
Molecular Weight (g/mol) 433.501
MDL Number MFCD00867568
InChI Key RTGDFNSFWBGLEC-SYZQJQIISA-N
Synonym mycophenolate mofetil, cellcept, munoloc, myfenax, mycophenolic acid morpholinoethyl ester, tm-mmf, mycophenylate mofetil, mycophenolatemofetil, cellcept tn, unii-9242ecw6r0
PubChem CID 5281078
ChEBI CHEBI:8764
IUPAC Name 2-morpholin-4-ylethyl (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoate
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)OCCN3CCOCC3)OC

Specifications

Melting Point 96°C
Color White
Quantity 1 g
Formula Weight 433.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Mycophenolate Mofetil
RTECSNumber MP7746700
TSCA No