missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolate Mofetil Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019444
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019444 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019444 Supplier Merck Emd Millipore Supplier No. PHR14511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Mycophenolate mofetil is a prodrug of mycophenolic acid that is utilized as an immunosuppressive agent for the treatment of renal transplant recipients. It may lead to gastrointestinal side effects.

Specifications

CAS 128794-94-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H31 NO7
MDL Number MFCD00867568
Synonym Mycophenolate mofetil; (4 E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid 2-(4-morpholinyl)ethyl ester; RS 61443; TM-MMF
Type Certified Reference Material
Quantity 1 g
Formula Weight 433.49
Grade Certified Reference Material