accessibility menu, dialog, popup

UserName

NH2ONHHO

CAS RN 127-07-1

IUPAC Name: hydroxyurea
Synonyms: hydroxyurea hydroxicarbamidum carbamohydroximic acid hydroxycarbamine ureaphil hydroxyurea (substance) hydroxycarbamid hydreia hydura hydrea
Molecular Weight (g/mol): 76.06
Molecular Formula: CH4N2O2
InChi Key: VSNHCAURESNICA-UHFFFAOYSA-N
SMILES: NC(=O)NO
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)