missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Hydroxyurea

Catalog No. 4000465GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4000465GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4000465GM Supplier MilliporeSigma™ Supplier No. 4000465GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase.

Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase. Induces apoptosis in chronic myelogenous leukemia lymphocytes. Reported to enhance the radiation effects in pancreatic adenocarcinoma cells. Releases NO in the presence of oxidizing agents.

Specifications

Color White
Description >98% by elemental analysis
Product Type Inhibitor
Target Proteins Inhibiting ribonucleotide reductase
CAS 127-07-1
Molecular Formula CH4N2O2
Formula Weight 76.1g/mol
Solubility H2O
Form Solid
Quantity 5 g