accessibility menu, dialog, popup

UserName

CH3NHNOHNSOONNF

CAS RN 1259028-99-3

IUPAC Name: 3-{4-[N-methyl1-(4-fluorophenyl)-1H-pyrazole-4-sulfonamido]phenyl}-1-phenylurea
Synonyms: 3-{4-[N-methyl1-(4-fluorophenyl)pyrazole-4-sulfonamido]phenyl}-1-phenylurea
Molecular Weight (g/mol): 465.5
Molecular Formula: C23H20FN5O3S
InChi Key: NSLIQOPYDUKWTA-UHFFFAOYSA-N
SMILES: CN(C1=CC=C(NC(=O)NC2=CC=CC=C2)C=C1)S(=O)(=O)C1=CN(N=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results