accessibility menu, dialog, popup

UserName

KCH3NClHONNNNN

CAS RN 124750-99-8

IUPAC Name: potassium 5-(4'-{[2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl]methyl}-[1,1'-biphenyl]-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Synonyms: potassium 5-(4'-{[2-butyl-4-chloro-5-(hydroxymethyl)imidazol-1-yl]methyl}-[1,1'-biphenyl]-2-yl)-1H-1,2,3,4-tetrazol-1-ide
Molecular Weight (g/mol): 461.01
Molecular Formula: C22H22ClKN6O
InChi Key: OXCMYAYHXIHQOA-UHFFFAOYSA-N
SMILES: [K+].CCCCC1=NC(Cl)=C(CO)N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NN=N[N-]1
Grade 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results