missing translation for 'onlineSavingsMsg'
Learn More

Losartan Potassium, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111012835
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012835 Supplier Merck Emd Millipore Supplier No. 61188100MG

Losartan potassium is an antihypertensive drug that is found to be an angiotensin II receptor type 1 blocker. It finds applications in reducing mechanical stress thereby having a control over the condition of cardiac hypertrophy.

Specifications

Packaging Glass Bottle
CAS 124750-99-8
Physical Form Powder/Crystals
Linear Formula C22H22ClKN6O
MDL Number MFCD02092704
Synonym 2-Butyl-4-chloro-1-{[2 '-(1 H-tetrazol-5-yl)(1,1 '-biphenyl)-4-yl]methyl}-1 H-imidazole-5-methanol monopotassium salt
Quantity 100 mg
Formula Weight 461.00
Grade Analytical Standard