missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Losartan Potassium 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Physical Form | Crystalline Powder |
| Quantity | 5 g |
| Formula Weight | 461.01 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | Losartan Potassium |
Safety and Handling
| RTECSNumber | NI6755100 |
| TSCA | No |