accessibility menu, dialog, popup

UserName

CAS RN 123-11-5

IUPAC Name: 4-methoxybenzaldehyde
Synonyms: anisaldehyde 18o-labeled 4-anisaldehyde 4-anisaldehyde, 18o-labeled aubepine obepin caswell no. 051E ccris 821 crategine 18O-labeled 4-anisaldehyde 4-anisaldehyde, 18O-labeled
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: ZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=C1
Percent Purity 
  • (4)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (14)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (5)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (3)
  • (2)
  • (3)
  • (4)
  • (7)
  • (4)
  • (3)
  • (3)
  • (4)
  • (14)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (5)
  • (4)
  • (2)
  • (3)