missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde 99.0+%, TCI America™

Catalog No. A0480500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0480500ML 500 mL
A048025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0480500ML Supplier TCI America Supplier No. A0480500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-11-5
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003385
InChI Key ZRSNZINYAWTAHE-UHFFFAOYSA-N
Synonym p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine
PubChem CID 31244
ChEBI CHEBI:28235
IUPAC Name 4-methoxybenzaldehyde
SMILES COC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Boiling Point 248°C
Quantity 500 mL
Formula Weight 136.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Anisaldehyde
EINECSNumber (3)-2661
RTECSNumber BZ2625000
TSCA Yes