missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde, 98%, Spectrum™ Chemical

Catalog No. 18612131
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612131 500 mL Poly Drum
18612130 5 mL Poly Drum
18612129 100 mL Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18612131 Supplier Spectrum Chemical Mfg Cor Supplier No. AN122500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-11-5

Specifications

CAS Max % 1
Boiling Point 248°C
Packaging Poly Drum
Quantity 500 mL
Percent Purity 0.98
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.