CAS RN 1229-55-6
IUPAC Name:
(1Z)-1-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms:
sudan red G
Molecular Weight (g/mol):
278.31
Molecular Formula:
C17H14N2O2
InChi Key:
BBBYRJLYBLPEBL-ZPHPHTNESA-N
SMILES:
COC1=CC=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12
1
–
2
of
2
results
1
Sudan red G, ≥96% (HPLC), MilliporeSigma™ Supelco™
Sudan dyes are classified under the azo dyes group of organic compounds that are commonly utilized as additive materials for industrial and scientific applications.
2
Sudan R 95.0+%, TCI America™
CAS: 1229-55-6 Molecular Formula: C17H14N2O2 Molecular Weight (g/mol): 278.31 MDL Number: MFCD00046377 InChI Key: BBBYRJLYBLPEBL-ZPHPHTNESA-N Synonym: 1-(2-Methoxyphenylazo)-2-naphthol, Solvent Red 1 PubChem CID: 5910713 IUPAC Name: (1Z)-1-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one SMILES: COC1=CC=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12