missing translation for 'onlineSavingsMsg'
Learn More

Sudan R 95.0+%, TCI America™

Catalog No. M048725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M048725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M048725G Supplier TCI America Supplier No. M048725G
Only null left

Chemical Identifiers

CAS 1229-55-6
Molecular Formula C17H14N2O2
Molecular Weight (g/mol) 278.31
MDL Number MFCD00046377
InChI Key BBBYRJLYBLPEBL-ZPHPHTNESA-N
Synonym 1-(2-Methoxyphenylazo)-2-naphthol, Solvent Red 1
PubChem CID 5910713
IUPAC Name (1Z)-1-[2-(2-methoxyphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES COC1=CC=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Specifications

Melting Point 179°C
Formula Weight 278.31
Percent Purity ≥95.0% (HPLC)
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Sudan R
EINECSNumber (5)-3080
RTECSNumber GE5844740
TSCA Yes