missing translation for 'onlineSavingsMsg'
Learn More

Sudan red G, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012812
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012812 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012812 Supplier Merck Emd Millipore Supplier No. 9128225MG

Sudan dyes are classified under the azo dyes group of organic compounds that are commonly utilized as additive materials for industrial and scientific applications.

Specifications

CAS 1229-55-6
Physical Form Neat
Linear Formula C17H14N2O2
MDL Number MFCD00046377
Synonym 1-(2-Methoxyphenylazo)-2-naphthol; Oil Red 113
Type Analytical Standard
Quantity 25 mg
Formula Weight 278.31
Percent Purity ≥96% (HPLC)
Grade Analytical Standard