CAS RN 12055-23-1
Hafnium(IV) oxide, 99.9% (metals basis excluding Zr), Zr <0.5%, Thermo Scientific Chemicals
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm, Thermo Scientific Chemicals
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium(IV) oxide, 98%, Thermo Scientific™
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium(IV) oxide, 99.99% (metals basis excluding Zr), Zr <100ppm, Thermo Scientific Chemicals
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium(IV) oxide, 99% (metals basis excluding Zr), Zr <1.5%, Thermo Scientific Chemicals
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium ICP Standard, 1000 ppm Hf in 3% HNO3, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
Hafnium AA Standard, 1000 ppm Hf in 3% HNO3/tr HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
VeriSpec™ Hafnium Standard for ICP 1000 ppm in 2% HCl/HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
VeriSpec™ Hafnium Standard for ICP/MS 100 ppm in 2% HNO3/HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO3/HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
VeriSpec™ Hafnium Standard for ICP 1000 ppm in 2% HNO3/HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]
VeriSpec™ Hafnium (Hf) Standard for AAS 1000 ppm in 2% HNO3/1% HF, Ricca Chemical
CAS: 12055-23-1 Molecular Formula: HfO2 Molecular Weight (g/mol): 210.49 MDL Number: MFCD00003565 InChI Key: WIHZLLGSGQNAGK-UHFFFAOYSA-N IUPAC Name: hafnium(4+) dioxidandiide SMILES: [O--].[O--].[Hf+4]