missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Hafnium Standard for ICP 1000 ppm in 2% HCl/HF, Ricca Chemical

Catalog No. RV010283500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010283500 500 mL
RV010283100 100 mL
RV010283250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010283500 Supplier Ricca Chemical Company Supplier No. RV010283500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HCl/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 12055-23-1 , 7647-01-0 , 7664-39-3 , 7732-18-5
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.43%, 1.96%, 0.49%, 0.12%
CAS Max % 99.33%, 2.04%, 0.51%, 0.12%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity