missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, 98%, Thermo Scientific™

Catalog No. AC192030010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192030010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC192030010 Supplier Thermo Scientific Chemicals Supplier No. 192030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12055-23-1
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Boiling Point 5400°C
Melting Point 2810°C
Quantity 1 g
Assay Percent Range 97.5% min
Linear Formula HfO2
Merck Index 10, 4470
Solubility Information Solubility in water: insoluble
Formula Weight 210.49
Percent Purity 98%
Infrared Spectrum White powder
Packaging Glass bottle
Specific Gravity 9.68
Density 9.6800g/mL
Chemical Name or Material Hafnium(IV) oxide, 98%
Show More Show Less
EINECSNumber 235-013-2

RUO – Research Use Only