missing translation for 'onlineSavingsMsg'
Learn More

Hafnium AA Standard, 1000 ppm Hf in 3% HNO3/tr HF, Ricca Chemical

Catalog No. p-7239393
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
AHF1KN100 Natural LDPE Bottle
AHF1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AHF1KN100 Supplier Ricca Chemical Company Supplier No. AHF1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12055-23-1 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.49, 0.11, 0.09
CAS Max % 97.0, 4.67, 0.11, 0.1
Odor Odorless
Concentration or Composition (by Analyte or Components) 1mL 1mg Hf (1, 000ppm Hf) Hf in 3% HNO3 / tr HF
Chemical Name or Material Hafnium AA Standard
ShelfLife 24 Months