missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium ICP Standard, 1000 ppm Hf in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 1,000ppm Hf |
| CAS Min % | 93.0, 4.49, 0.11, 0.09 |
| CAS Max % | 97.0, 4.67, 0.11, 0.1 |
| Equivalence | 1 mL = 1 mg Hf |
| Chemical Name or Material | Hafnium ICP Standard |