CAS RN 12027-06-4
Ammonium iodide, 99+%, for analysis
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium iodide, 99.995% (metals basis)
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium iodide, ACS, 99.0% min
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium iodide, 99%, extra pure
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium iodide, 98+%
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium iodide, ACS reagent
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium Iodide, (ACS Reagent Grade), MP Biomedicals
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: ammonium iodide SMILES: [NH4+].[I-]
Ammonium Iodide, puriss. p.a. ACS Reagent, ≥99.0% (AT), Solstice
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 IUPAC Name: azanium;iodide SMILES: [NH4+].[I-]
Ammonium Iodide, ACS, 99%, Spectrum™ Chemical
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N IUPAC Name: ammonium iodide SMILES: [NH4+].[I-]
Ammonium Iodide, puriss., Solstice
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 MDL Number: MFCD00011424 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N Synonym: ammonium iodide,azanium iodide,unii-oz8f027ldh,ammonium iodide nh4 i,oz8f027ldh,ammonium-iodid,ammonium jodatum,ammoniumiodid,hsdb 476,nh4i PubChem CID: 25487 ChEBI: CHEBI:63040 SMILES: [NH4+].[I-]
Ammonium Iodide, ACS Reagent, Reagents
CAS: 12027-06-4 Molecular Formula: H4IN Molecular Weight (g/mol): 144.94 InChI Key: UKFWSNCTAHXBQN-UHFFFAOYSA-N IUPAC Name: ammonium iodide SMILES: [NH4+].[I-]