missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iodide, ACS, 99.0% min
Description
Appropriately manufactured ammonium iodide is used in pharmaceutical industries. It is used in photographic chemicals, some medications and several chemical industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 551°C (sublimation) |
| Color | White to Yellow |
| Physical Form | Granules |
| Quantity | 500 g |
| Odor | Odorless |
| Refractive Index | 1.7031 |
| Sensitivity | Air and light sensitive; Hygroscopic |
| Assay Percent Range | 99.0% min |
| Density | 2.514 g/mL |
| Merck Index | 14,529 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 234-717-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only