missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, puriss. p.a. ACS Reagent, ≥99.0% (AT), Solstice

Catalog No. 60044626
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044626 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044626 Supplier Solstice Supplier No. 09874100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., ACS reagent,≥99.0% (AT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Color White
pH 4.5 to 6.5 (at 20°C)
Quantity 100 g
Linear Formula NH4I
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 50mg/mL
Formula Weight 144.94g/mol
Percent Purity ≥99.0% (AT)
Grade ACS/puriss. p.a.
Packaging Glass Bottle
Residue after Ignition ≤0.02% (as SO4)
Appearance Crystals
Vapor Pressure 1mmHg (210.9°C)
Density 2.510g/cu.cm
Chemical Name or Material Ammonium Iodide
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 234-717-7