missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iodide, ACS, 99%, Spectrum™ Chemical

Catalog No. 18608839
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608839 50 kg Amber Glass Bottle
18608836 125 g Amber Glass Bottle
18608838 500 g Amber Glass Bottle
18608693 2.5 kg Amber Glass Bottle
18608837 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608839 Supplier Spectrum Chemical Mfg Cor Supplier No. A120550KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
IUPAC Name ammonium iodide
SMILES [NH4+].[I-]

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.0005
Chemical Name or Material Ammonium Iodide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.