missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iodide, (ACS Reagent Grade), MP Biomedicals
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 500 g |
| Formula Weight | 144.9 |
| Grade | ACS Reagent |
| Chemical Name or Material | Ammonium Iodide |
Safety and Handling
| Recommended Storage | Room Temperature |