missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iodide, 98+%
Description
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.
Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.
Notes
Store at 4°C. Hande under dry protective gas.
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.
Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.
Notes
Store at 4°C. Hande under dry protective gas.
Specifications
Specifications
| Melting Point | 551°C (sublimation) |
| Quantity | 500 g |
| Odor | Odorless |
| Refractive Index | 1.7031 |
| Sensitivity | Air and light sensitive; Hygroscopic |
| Density | 2.514 |
| Merck Index | 14,529 |
| Solubility Information | Soluble in water (1770mg/ml at 25°C),methanol (400mg/ml),glycerol (667mg/ml),alcohol (270mg/ml),and ethanol. |
| Formula Weight | 144.94 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 234-717-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only