accessibility menu, dialog, popup

UserName

ClPdClPdH2CH2CCH2CH2

CAS RN 12012-95-2

IUPAC Name: Allylpalladium chloride dimer
Synonyms: —
Molecular Weight (g/mol): 365.89
Molecular Formula: C6H10Cl2Pd2
InChi Key: PENAXHPKEVTBLF-UHFFFAOYSA-L
SMILES: Cl[Pd].Cl[Pd].c:c:c.c:c:c
Physical Form 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)