missing translation for 'onlineSavingsMsg'
Learn More

Allylpalladium(II) Chloride Dimer 97.0+%, TCI America™

Catalog No. A1479500MG
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1479500MG 500 mg
A14791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1479500MG Supplier TCI America Supplier No. A1479500MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12012-95-2
Molecular Formula C6H10Cl2Pd2
Molecular Weight (g/mol) 365.89
MDL Number MFCD00044874
InChI Key PENAXHPKEVTBLF-UHFFFAOYSA-L
Synonym allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
PubChem CID 61538
IUPAC Name Allylpalladium chloride dimer
SMILES Cl[Pd].Cl[Pd].c:c:c.c:c:c

Specifications

Quantity 500 mg
Formula Weight 365.89
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Allylpalladium(II) Chloride Dimer
RTECSNumber RT3510000
TSCA Yes