accessibility menu, dialog, popup

UserName

CH3OONH

CAS RN 1193-62-0

IUPAC Name: methyl 1H-pyrrole-2-carboxylate
Synonyms: methyl pyrrole-2-carboxylate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: VONGYFFEWFJHNP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)