missing translation for 'onlineSavingsMsg'
Learn More

Methyl Pyrrole-2-carboxylate 98.0+%, TCI America™

Catalog No. M21605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21605G 5 g
M216025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21605G Supplier TCI America Supplier No. M21605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-62-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00817048
InChI Key VONGYFFEWFJHNP-UHFFFAOYSA-N
Synonym methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1
PubChem CID 136930
IUPAC Name methyl 1H-pyrrole-2-carboxylate
SMILES COC(=O)C1=CC=CN1

Specifications

Melting Point 73°C
Quantity 5 g
Formula Weight 125.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Pyrrole-2-carboxylate