Learn More
Methyl pyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals
Description
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Specifications
Specifications
| Melting Point | 74°C to 78°C |
| Quantity | 5 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 125.13 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl pyrrole-2-carboxylate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only