accessibility menu, dialog, popup

UserName

CAS RN 117068-71-0

ONBr

CAS RN 117068-71-0

IUPAC Name: 2-(benzyloxy)-6-bromopyridine
Synonyms: 2-(benzyloxy)-6-bromopyridine
Molecular Weight (g/mol): 264.12
Molecular Formula: C12H10BrNO
InChi Key: WEMPEMXDSUATEK-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OCC2=CC=CC=C2)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results