missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-benzyloxypyridine 97.0+%, TCI America™

Catalog No. B25765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B25765G 5 g
B257625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25765G Supplier TCI America Supplier No. B25765G
Only null left

Chemical Identifiers

CAS 117068-71-0
Molecular Formula C12H10BrNO
Molecular Weight (g/mol) 264.12
MDL Number MFCD00234782
InChI Key WEMPEMXDSUATEK-UHFFFAOYSA-N
PubChem CID 817475
IUPAC Name 2-(benzyloxy)-6-bromopyridine
SMILES BrC1=CC=CC(OCC2=CC=CC=C2)=N1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 264.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-6-benzyloxypyridine
TSCA No
Recommended Storage Refrigerator