accessibility menu, dialog, popup

UserName

H3COOH3COCH3CH3O

CAS RN 1136-86-3

IUPAC Name: 1-(3,4,5-trimethoxyphenyl)ethan-1-one
Synonyms: 1-(3,4,5-trimethoxyphenyl)ethanone
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: VUGQIIQFXCXZJU-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)