accessibility menu, dialog, popup

UserName

H3COOCH3NNSH3COO

CAS RN 113583-35-0

IUPAC Name: 2-methanesulfonyl-4,6-dimethoxypyrimidine
Synonyms: 2-methanesulfonyl-4,6-dimethoxypyrimidine
Molecular Weight (g/mol): 218.23
Molecular Formula: C7H10N2O4S
InChi Key: ITDVJJVNAASTRS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(=N1)S(C)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results