missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine 98.0+%, TCI America™

Catalog No. D31015G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31015G Supplier TCI America Supplier No. D31015G

Chemical Identifiers

CAS 113583-35-0
Molecular Formula C7H10N2O4S
Molecular Weight (g/mol) 218.23
MDL Number MFCD00672151
InChI Key ITDVJJVNAASTRS-UHFFFAOYSA-N
Synonym 4,6-dimethoxy-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfony-4,6-dimethoxypyridine, 4,6-dimethoxy-2-methylsulfonyl-pyrimidine, 2-methanesulfonyl-4,6-dimethoxy-pyrimidine, 4,6-dimethoxy-2-methylsulphonyl pyrimidine, pyrimidine, 4,6-dimethoxy-2-methylsulfonyl, pubchem23351, maybridge1_008822
PubChem CID 838363
IUPAC Name 2-methanesulfonyl-4,6-dimethoxypyrimidine
SMILES COC1=CC(OC)=NC(=N1)S(C)(=O)=O

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 218.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine