accessibility menu, dialog, popup

UserName

CH3OOSOONH2

CAS RN 112941-26-1

IUPAC Name: methyl 2-(sulfamoylmethyl)benzoate
Synonyms: methyl 2-(sulfamoylmethyl)benzoate
Molecular Weight (g/mol): 229.25
Molecular Formula: C9H11NO4S
InChi Key: DBOUFTHAEAVMJC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1CS(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results