missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Aminosulfonylmethyl)benzoate 98.0+%, TCI America™

Catalog No. M186725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M186725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M186725G Supplier TCI America Supplier No. M186725G

Chemical Identifiers

CAS 112941-26-1
Molecular Formula C9H11NO4S
Molecular Weight (g/mol) 229.25
MDL Number MFCD03265370
InChI Key DBOUFTHAEAVMJC-UHFFFAOYSA-N
Synonym 2-(Aminosulfonylmethyl)benzoic Acid Methyl Ester, o-Carbomethoxybenzyl Sulfonamide
PubChem CID 13958774
IUPAC Name methyl 2-(sulfamoylmethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1CS(=O)(=O)N

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 25 g
Formula Weight 229.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(Aminosulfonylmethyl)benzoate