accessibility menu, dialog, popup

UserName

NNNNN

CAS RN 112809-51-5

IUPAC Name: 4-[(4-cyanophenyl)(1H-1,2,4-triazol-1-yl)methyl]benzonitrile
Synonyms: letrozole fémara femara 来曲唑 LTZ letrozole preparation letrozol novartis brand of letrozole letrozole (product) letrozole (substance)
Molecular Weight (g/mol): 285.31
Molecular Formula: C17H11N5
InChi Key: HPJKCIUCZWXJDR-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C(N1C=NC=N1)C1=CC=C(C=C1)C#N
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results