missing translation for 'onlineSavingsMsg'
Learn More

Letrozole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012510
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012510 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012510 Supplier Merck Emd Millipore Supplier No. PHR15401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Letrozole is used for the treatment of breast cancer in hormone-dependent postmenopausal patients. The drug works by blocking the synthesis of estrogen, a causative agent for cancer.

Specifications

CAS 112809-51-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H11N5
MDL Number MFCD00866241
Synonym Letrozole; 4,4′-(1 H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile
Type Certified Reference Material
Quantity 1 g
Formula Weight 285.30
Grade Certified Reference Material