missing translation for 'onlineSavingsMsg'
Learn More

Letrozole 98.0+%, TCI America™

Catalog No. L02481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L02481G Supplier TCI America Supplier No. L02481G

Chemical Identifiers

CAS 112809-51-5
Molecular Formula C17H11N5
Molecular Weight (g/mol) 285.31
MDL Number MFCD00866241
InChI Key HPJKCIUCZWXJDR-UHFFFAOYSA-N
Synonym letrozole, femara, 4,4'-1h-1,2,4-triazol-1-yl methylene dibenzonitrile, letrozol, letoval, letrazole, femera, letrozole usan:inn, unii-7lkk855w8i, 4,4'-1h-1,2,4-triazol-1-ylmethanediyl dibenzonitrile
PubChem CID 3902
ChEBI CHEBI:6413
IUPAC Name 4-[(4-cyanophenyl)-(1,2,4-triazol-1-yl)methyl]benzonitrile
SMILES C1=CC(=CC=C1C#N)C(C2=CC=C(C=C2)C#N)N3C=NC=N3

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 285.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Letrozole
RTECSNumber DI4957000
TSCA No
Recommended Storage Refrigerator