accessibility menu, dialog, popup

UserName

CAS RN 112529-15-4

HClH3COSOHNON

CAS RN 112529-15-4

IUPAC Name: hydrogen 5-({4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl}methyl)-1,3-thiazolidine-2,4-dione chloride
Synonyms: hydrogen pioglitazone chloride
Molecular Weight (g/mol): 392.9
Molecular Formula: C19H21ClN2O3S
InChi Key: GHUUBYQTCDQWRA-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCC1=CC=C(CCOC2=CC=C(CC3SC(=O)NC3=O)C=C2)N=C1
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)